Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyltrimethoxysilane, 97%, AcroSeal™

Catalog No. AC433081000
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC433081000 100 mL AcroSeal™ Glass Bottle
AC433080010 1 L AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC433081000 Supplier Thermo Scientific Chemicals Supplier No. 433081000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5314-55-6 | C5H14O3Si | 150.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5314-55-6
Molecular Formula C5H14O3Si
Molecular Weight (g/mol) 150.25
InChI Key SBRXLTRZCJVAPH-UHFFFAOYSA-N
Synonym silane, ethyltrimethoxy, ethyl trimethoxy silane, trimethoxyethylsilane, silane,ethyltrimethoxy, acmc-209l4b, ethyltrimethoxysilane, dow corning™ product 1-6321
PubChem CID 79204
IUPAC Name ethyl(trimethoxy)silane
SMILES CC[Si](OC)(OC)OC

Specifications

Color Colorless to Yellow
Density 0.9400g/mL
Flash Point 27°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.3830 to 1.3850 (20°C, 589nm)
Quantity 100 mL
Specific Gravity 0.94
Solubility Information Solubility in water: decomposes
Formula Weight 150.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyltrimethoxysilane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 226-172-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.