Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyltrimethylammonium Iodide 98.0+%, TCI America™
SDP

Catalog No. E019025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E019025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E019025G Supplier TCI America Supplier No. E019025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 51-93-4
Molecular Formula C5H14IN
Molecular Weight (g/mol) 215.08
MDL Number MFCD00054229
InChI Key ZPEBBUBSCOELHI-UHFFFAOYSA-M
PubChem CID 66125
ChEBI CHEBI:55320
IUPAC Name ethyltrimethylazanium iodide
SMILES [I-].CC[N+](C)(C)C

Specifications

Color White
Quantity 25 g
Formula Weight 215.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyltrimethylammonium Iodide
EINECSNumber (2)-0186
RTECSNumber BQ6045000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.