Learn More
Medchemexpress LLC Ethylvanillin | 121-32-4 | 99.9% | 250 MG
SDP
Catalog No. 5000405580
Encompass_Preferred
Ethylvanillin is an orally active antioxidant capable of influencing cytochrome P450 activity in rats. It also serves as a food flavoring agent in various food processing applications.
  • Inhibits AAPH-induced hemolysis in erythrocyte suspension
  • Shows antioxidant activity in erythrocyte suspension in ABTS radical assay
  • Increases plasma ethyl vanillic acid levels and antioxidant activity in mice
  • Induces CYP2E1 activity and inhibits CYP1A2 activity in rats
Catalog No. 50-004-05580 Supplier Medchemexpress LLC Supplier No. 250MGHYB0940
Ethylvanillin is an orally active antioxidant capable of influencing cytochrome P450 activity in rats. It also serves as a food flavoring agent in various food processing applications.
  • Inhibits AAPH-induced hemolysis in erythrocyte suspension
  • Shows antioxidant activity in erythrocyte suspension in ABTS radical assay
  • Increases plasma ethyl vanillic acid levels and antioxidant activity in mice
  • Induces CYP2E1 activity and inhibits CYP1A2 activity in rats