Learn More
Medchemexpress LLC Ethylvanillin | 121-32-4 | 99.9% | 500 MG
SDP
Catalog No. 5000407912
Encompass_Preferred
Ethylvanillin is an orally active antioxidant that can affect cytochrome P450 activity in rats. It can also be used as a food flavoring agent in the field of food processing.
  • Exhibits antioxidant activity in erythrocyte suspension and in mice.
  • Affects cytochrome P450 activity, inducing CYP2E1 and inhibiting CYP1A2 in rats.
  • Soluble in ethanol, DMSO, and water.
  • Inhibits AAPH-induced hemolysis.
  • Increases plasma ethyl vanillic acid levels and antioxidant activity in ICR mice.
Catalog No. 50-004-07912 Supplier Medchemexpress LLC Supplier No. HYB0940500MG
Ethylvanillin is an orally active antioxidant that can affect cytochrome P450 activity in rats. It can also be used as a food flavoring agent in the field of food processing.
  • Exhibits antioxidant activity in erythrocyte suspension and in mice.
  • Affects cytochrome P450 activity, inducing CYP2E1 and inhibiting CYP1A2 in rats.
  • Soluble in ethanol, DMSO, and water.
  • Inhibits AAPH-induced hemolysis.
  • Increases plasma ethyl vanillic acid levels and antioxidant activity in ICR mice.