Learn More
Apexbio Technology LLC ETHYLVANILLIN-200MG
SDP
Catalog No. 5000570012
Encompass_Preferred
Ethylvanillin (CAS No 121-32-4) is a synthetic flavor compound primarily used in the food industry but has gained attention in biomedical research Structurally related to vanillin it is investigated for its antioxidant and antimicrobial properties Mechanistically ethylvanillin may interact with pathways involved in oxidative stress reduction though specific targets are still under investigation In vitro studies often demonstrate activity in the nanomolar to low micromolar range highlighting its potential efficacy Common research applications include its use in various cell models to assess cytoprotective effects and in animal studies to explore its therapeutic potential Additionally it is utilized in drug screening programs to evaluate its additive or synergistic effects with other compounds Experimental concentrations typically depend on the specific research design adapting to the particular cell type or model system employed in the study
Catalog No. 50-005-70012 Supplier Apexbio Technology LLC Supplier No. C6766200
Ethylvanillin (CAS No 121-32-4) is a synthetic flavor compound primarily used in the food industry but has gained attention in biomedical research Structurally related to vanillin it is investigated for its antioxidant and antimicrobial properties Mechanistically ethylvanillin may interact with pathways involved in oxidative stress reduction though specific targets are still under investigation In vitro studies often demonstrate activity in the nanomolar to low micromolar range highlighting its potential efficacy Common research applications include its use in various cell models to assess cytoprotective effects and in animal studies to explore its therapeutic potential Additionally it is utilized in drug screening programs to evaluate its additive or synergistic effects with other compounds Experimental concentrations typically depend on the specific research design adapting to the particular cell type or model system employed in the study