Learn More
Medchemexpress LLC Ethylvanillin-d5 | 1335401-74-5 | 98.0% | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0940S10MG
Encompass_Preferred
Ethylvanillin-d5 is a deuterium-labeled ethylvanillin supplied as a solid reference standard for analytical and research use, commonly applied in mass spectrometry and tracer studies. It provides a stable isotopic internal standard for quantitation and method validation.
  • Deuterium-labeled (d5) ethylvanillin for use as an isotope standard.
  • Suitable as an internal standard for LC-MS/MS and GC-MS assays.
  • Provided as a solid powder for analytical workflows.
  • High isotopic enrichment and analytical-grade purity for reference use.
Catalog No. 50-002-96741