Learn More
Rieke Metals LLC ETHYLZINC BROMIDE 100G
Catalog No. NC3522061
Encompass
Formula: C2H5BrZnMolecular Weight: 174.3555CAS: 6107-37-5Notes: 0.50 M in THFMDL: MFCD22685696
Catalog No. NC3522061 Supplier Rieke Metals LLC Supplier No. ZT1274100ML
Formula: C2H5BrZnMolecular Weight: 174.3555CAS: 6107-37-5Notes: 0.50 M in THFMDL: MFCD22685696