Learn More
Apexbio Technology LLC Ethynodiol diacetate 297-76-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000090510
Encompass_Preferred
Ethynodiol diacetate (297-76-7) is a small-molecule agonist targeting progesterone receptors It is designed to selectively bind and activate these receptors thereby modulating gene transcription pathways and influencing reproductive tissue responses Ethynodiol diacetate exerts its biological activity primarily through selective activation of progesterone receptors Based on these pharmacological properties Ethynodiol diacetate holds research potential in studies investigating hormonal regulation contraceptive mechanisms and modulation of the reproductive system
Catalog No. 50-000-90510 Supplier Apexbio Technology LLC Supplier No. B15085.1
Ethynodiol diacetate (297-76-7) is a small-molecule agonist targeting progesterone receptors It is designed to selectively bind and activate these receptors thereby modulating gene transcription pathways and influencing reproductive tissue responses Ethynodiol diacetate exerts its biological activity primarily through selective activation of progesterone receptors Based on these pharmacological properties Ethynodiol diacetate holds research potential in studies investigating hormonal regulation contraceptive mechanisms and modulation of the reproductive system