Learn More
Medchemexpress LLC Ethynodiol diacetate | 297-76-7 | 99.9% | 1 ML
SDP
Catalog No. 5000414023
Encompass_Preferred
Ethynodiol diacetate is a click chemistry reagent that contains an Alkyne group. It can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
Features and benefits:
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with molecules containing azide groups
Catalog No. 50-004-14023 Supplier Medchemexpress LLC Supplier No. HYB108910MM/1ML
Ethynodiol diacetate is a click chemistry reagent that contains an Alkyne group. It can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
Features and benefits:
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with molecules containing azide groups