Learn More
Medchemexpress LLC Ethynodiol diacetate | 297-76-7 | MFCD00083326 | 99.85% | 1 G
SDP
Catalog No. 5000413868
Encompass_Preferred
Ethynodiol diacetate is a click chemistry reagent that contains an Alkyne group. It can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. It is for research use only and not sold to patients.
  • Click chemistry reagent
  • Contains an Alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-004-13868 Supplier Medchemexpress LLC Supplier No. HYB10891G
Ethynodiol diacetate is a click chemistry reagent that contains an Alkyne group. It can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. It is for research use only and not sold to patients.
  • Click chemistry reagent
  • Contains an Alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups