Learn More
Cayman Chemical Ethynodiol Diacetate, Purity: ≥98%, CAS: 297-76-7, 500 mg
SDP
Catalog No. 502978721
Encompass_Preferred
A synthetic progestogen; prevents the development of osteoporosis in ovariectomized rats at 9 µg/animal per day; inhibits copulatory ovulation in rabbits
Catalog No. 50-297-8721 Supplier Cayman Chemical Supplier No. 29854500 MG
A synthetic progestogen; prevents the development of osteoporosis in ovariectomized rats at 9 µg/animal per day; inhibits copulatory ovulation in rabbits