Learn More
Selleck Chemical LLC Ethynodiol diacetate S3160-50mg
SDP
Catalog No. 503109221
Encompass_Preferred
Ethynodiol diacetate (8080 CB Ethynodiol acetate) is one of the first synthetic progestogens used in contraceptive pills
Catalog No. 50-310-9221 Supplier Selleck Chemical LLC Supplier No. S316050MG
Ethynodiol diacetate (8080 CB Ethynodiol acetate) is one of the first synthetic progestogens used in contraceptive pills