Learn More
Cayman Chemical Ethynyl Estradiol
SDP
Catalog No. 502889745
Encompass_Preferred
An estrogen receptor agonist (EC50 = 8.5 pM in a reporter assay using T47D cells); reduces serum cholesterol levels and femur bone mass and increases uterine mass in an OVX rat model of postmenopause at 0.1 mg/kg per day; has been found in wastewater effluent; prevents secondary sexual development in male fathead minnows and increases the numbers of intersex fish at 4 ng/l
Catalog No. 50-288-9745 Supplier Cayman Chemical Supplier No. 100064861 G
Add to Cart
Add to Cart
An estrogen receptor agonist (EC50 = 8.5 pM in a reporter assay using T47D cells); reduces serum cholesterol levels and femur bone mass and increases uterine mass in an OVX rat model of postmenopause at 0.1 mg/kg per day; has been found in wastewater effluent; prevents secondary sexual development in male fathead minnows and increases the numbers of intersex fish at 4 ng/l