Learn More
Medchemexpress LLC Ethynyl Estradiol-d4 | 350820-06-3 | 99.0% | 5 MG
SDP
Catalog No. 5000406909
Encompass_Preferred
Ethynyl Estradiol-d4 is the deuterium labeled Ethynyl Estradiol. It is an orally bio-active estrogen used in almost all modern formulations of combined oral contraceptive pills. This compound is a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-06909 Supplier Medchemexpress LLC Supplier No. HYB0216S5MG
Ethynyl Estradiol-d4 is the deuterium labeled Ethynyl Estradiol. It is an orally bio-active estrogen used in almost all modern formulations of combined oral contraceptive pills. This compound is a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.