Learn More
Cayman Chemical EthynylcytIdIn 10mg
SDP
Catalog No. 502938215
Encompass_Preferred
A nucleoside analog with anticancer activity; cytotoxic to A549, NUGC-3, HT-29, MIA PaCa-2, and MCF-7 cancer cells (IC50s = 0.114, 1.032, 0.539, 0.198, and 0.326 µM, respectively); reduces tumor growth in CADO-LC11 lung, PAN-12 pancreas, AZ-521 stomach, and CO-3 colon cancer mouse xenograft models at 0.25 mg/kg per day
Catalog No. 50-293-8215 Supplier Cayman Chemical Supplier No. 2768110 MG
A nucleoside analog with anticancer activity; cytotoxic to A549, NUGC-3, HT-29, MIA PaCa-2, and MCF-7 cancer cells (IC50s = 0.114, 1.032, 0.539, 0.198, and 0.326 µM, respectively); reduces tumor growth in CADO-LC11 lung, PAN-12 pancreas, AZ-521 stomach, and CO-3 colon cancer mouse xenograft models at 0.25 mg/kg per day