Learn More
Medchemexpress LLC Ethynylcytidine | 180300-43-0 | 100.0% | 267.24 | 5 MG
SDP
Catalog No. 5000394836
Encompass_Preferred
Ethynylcytidine (ECyD) is a nucleoside analog and a potent inhibitor of RNA synthesis, inhibiting RNA polymerases I, II, and III. It exhibits robust antitumor activity in a wide range of cancer models. Additionally, Ethynylcytidine functions as a click chemistry reagent, as it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Potent inhibitor of RNA synthesis
  • Inhibits RNA polymerases I, II, and III
  • Exhibits robust antitumor activity in various cancer models
  • Functions as a click chemistry reagent
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules
Catalog No. 50-003-94836 Supplier Medchemexpress LLC Supplier No. HY162005MG
Ethynylcytidine (ECyD) is a nucleoside analog and a potent inhibitor of RNA synthesis, inhibiting RNA polymerases I, II, and III. It exhibits robust antitumor activity in a wide range of cancer models. Additionally, Ethynylcytidine functions as a click chemistry reagent, as it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Potent inhibitor of RNA synthesis
  • Inhibits RNA polymerases I, II, and III
  • Exhibits robust antitumor activity in various cancer models
  • Functions as a click chemistry reagent
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules