Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethynylferrocene

Catalog No. 4426581GA Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-265-81GA 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-265-81GA Supplier Sigma Aldrich Supplier No. 4426581GA
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethynylferrocene

Specifications

CAS 1271-47-2
Melting Point 105°C to 110°C
Molecular Formula C7H9BO3
Linear Formula CH3OC6H4B(OH)2
Quantity 5 g
MDL Number MFCD00236047
Synonym (2-Methoxyphenyl)boric acid; 2-Methoxybenzeneboronic acid; o-Anisylboronic acid; o-Methoxybenzeneboronic acid; o-Methoxyphenylboronic acid
Molecular Weight (g/mol) 151.96 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.