Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethynylmagnesium Chloride Solution

Catalog No. 346160800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-616-0800ML 800 mL
34-616-0100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 34-616-0800ML Supplier Sigma Aldrich Supplier No. 346160800ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethynylmagnesium chloride solution can be used:To synthesize 1-ethynyldeoxyribose derivatives which can further undergo Sonogashira coupling with various aryl, heteroaryl, and metallocenyl derivatives.In one of the key synthetic steps of substituted naphthostyrils.In the preparation of propargylic ester precursor for the synthesis of 1,3-dienylphosphonates.

Specifications

CAS 65032-27-1
Density 0.921 g/mL at 25°C
Molecular Formula C2HClMg
Linear Formula CH///CMgCl
MDL Number MFCD00075343
Quantity 800 mL
Molecular Weight (g/mol) 84.79 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.