Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethynyltrimethylsilane

Catalog No. 2181705G
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-817-05G 5 g
50-289-8583 25 g
21-817-01G 1 g
21-817-0100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 21-817-05G Supplier Sigma Aldrich Supplier No. 2181705G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Laser-induced polymerization of gaseous ethynyltrimethylsilane was used for efficient chemical vapour deposition of polycarbosilane films. ethynyltrimethylsilane acts as substrate for nickel-catalyzed cross-coupling with benzonitriles.ethynyltrimethylsilane was used in:
  • Microwave-assisted, one-pot, three-step sonogashira cross-coupling-desilylation-cycloaddition reaction for the preparation of 1,4-disubstituted 1,2,3-triazoles
  • Synthesis of poly(ethynyltrimethylsilane) containing pd (ii) coordination sites
  • Pyrazole synthesis via 1,3-dipolar cycloaddition of diazo compounds to acetylenes

Specifications

CAS 1066-54-2
Density 0.709 g/cm3 (literature)
Boiling Point 53°C (lit.)
Molecular Formula C5H10Si
Refractive Index n20/D 1.388 (literature)
Linear Formula (CH3)3 SiC///CH
MDL Number MFCD00008569
Quantity 5 g
Synonym Trimethylsilylacetylene
Molecular Weight (g/mol) 98.22
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.