Learn More
Medchemexpress LLC Eticlopride hydrochloride | 97612-24-3 | MFCD00055132 | 99.5% | 377.31 g/mol | C17H26Cl2N2O3 | 1 ML
SDP
Catalog No. 5000246554
Encompass_Preferred
Eticlopride hydrochloride is a selective dopamine D2-like receptor antagonist used in pharmacology and neuroscience research. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and is also available in solid form for assay development and receptor binding studies.
  • Selective D2 receptor antagonist with high affinity
  • Available as 10 mM solution in DMSO (1 mL) or as solid quantities
  • High reported purity suitable for research applications
  • Used for receptor pharmacology and binding assays
  • Identified by CAS number for traceability
Catalog No. 50-002-46554 Supplier Medchemexpress LLC Supplier No. HY10341310MM 1ML
Eticlopride hydrochloride is a selective dopamine D2-like receptor antagonist used in pharmacology and neuroscience research. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and is also available in solid form for assay development and receptor binding studies.
  • Selective D2 receptor antagonist with high affinity
  • Available as 10 mM solution in DMSO (1 mL) or as solid quantities
  • High reported purity suitable for research applications
  • Used for receptor pharmacology and binding assays
  • Identified by CAS number for traceability