Learn More
TARGETMOL CHEMICALS INC Etidocaine Hydrochloride 1MG
SDP
Catalog No. 502876803
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes.Etidocaine Hydrochloride (W19053) is a long-acting anesthetic and a blocker of the voltage-gated sodium channel. Purity 98.71%
Catalog No. 50-287-6803 Supplier TARGETMOL CHEMICALS INC Supplier No. T317161MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes.Etidocaine Hydrochloride (W19053) is a long-acting anesthetic and a blocker of the voltage-gated sodium channel. Purity 98.71%