Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Etidocaine hydrochloride | 36637-19-1 | 99.9% | 100 MG
SDP

Catalog No. 5000410741
Encompass_Preferred

Etidocaine hydrochloride is a long-acting aminoamide local anesthetic agent used in research to suppress or relieve pain. It is intended for research use only and not for human or animal therapeutic use.

  • Long-acting aminoamide local anesthetic agent
  • Induces a significant increase in human fibroblasts survival in IGL-EDC formulations
  • Did not show postinjection neurologic deficit when administered as a spinal injection at 0.0075% in adult Swiss Webster male mice
  • Purity of 99.9%
  • Molecular weight of 312.88
  • Chemical formula C17H29ClN2O
  • Appears as a solid
  • White to off-white color
  • Soluble in DMSO at 62.5 mg/mL

Catalog No. 50-004-10741 Supplier Medchemexpress LLC Supplier No. HYB2080A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Etidocaine hydrochloride is a long-acting aminoamide local anesthetic agent used in research to suppress or relieve pain. It is intended for research use only and not for human or animal therapeutic use.

  • Long-acting aminoamide local anesthetic agent
  • Induces a significant increase in human fibroblasts survival in IGL-EDC formulations
  • Did not show postinjection neurologic deficit when administered as a spinal injection at 0.0075% in adult Swiss Webster male mice
  • Purity of 99.9%
  • Molecular weight of 312.88
  • Chemical formula C17H29ClN2O
  • Appears as a solid
  • White to off-white color
  • Soluble in DMSO at 62.5 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.