Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Etidronate disodium

Catalog No. AAJ6238103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6238103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6238103 Supplier Thermo Scientific Chemicals Supplier No. J6238103
  • $327.00
    $277.65 / Each of 1

    Save $49.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7414-83-7 | C2H6Na2O7P2 | 249.99 g/mol

Etidronate disodium is used as a bisphosphonate bone resorption inhibitor. Etidronic acid is used as a calcium regulator. Etidronic acid is a bisphosphonate antiresorptive agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Etidronate disodium is used as a bisphosphonate bone resorption inhibitor. Etidronic acid is used as a calcium regulator. Etidronic acid is a bisphosphonate antiresorptive agent.

Solubility
Soluble in water (26 mg/m
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7414-83-7
Molecular Formula C2H6Na2O7P2
Molecular Weight (g/mol) 249.99
MDL Number MFCD00152567
InChI Key GWBBVOVXJZATQQ-UHFFFAOYSA-L
Synonym etidronate disodium, disodium etidronate, didronel, sodium etidronate, sodium ethidronate, sodium ethydronate, disodium ethydronate, didronel r, disodium ethanol-1,1-diphosphonate, etidronsaeure dinatriumsalz
PubChem CID 23894
ChEBI CHEBI:4906
IUPAC Name disodium hydrogen (1-hydrogen phosphonato-1-hydroxyethyl)phosphonate
SMILES [Na+].[Na+].CC(O)(P(O)([O-])=O)P(O)([O-])=O

Specifications

Melting Point >300°C
Color White
Quantity 1 g
Merck Index 14,3863
Solubility Information Soluble in water (26mg/ml).
Formula Weight 249.99
Physical Form Crystalline Solid
Chemical Name or Material Etidronate disodium
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
EINECSNumber 231-025-7
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.