Learn More
Cayman Chemical Etidronate (sodium salt), Purity: ≥95%, CAS: 7414-83-7, 1 g
SDP
Catalog No. 502984906
Encompass_Preferred
A non-nitrogenous bisphosphonate that binds to hydroxyapatite and reduces osteoclast proliferation, thereby inhibiting abnormal bone resorption
Catalog No. 50-298-4906 Supplier Cayman Chemical Supplier No. 207691 G
A non-nitrogenous bisphosphonate that binds to hydroxyapatite and reduces osteoclast proliferation, thereby inhibiting abnormal bone resorption