Learn More
Cayman Chemical Etidronate (sodium salt), Purity: ≥95%, CAS: 7414-83-7, 500 mg
SDP
Catalog No. 502982736
Encompass_Preferred
A non-nitrogenous bisphosphonate that binds to hydroxyapatite and reduces osteoclast proliferation, thereby inhibiting abnormal bone resorption
Catalog No. 50-298-2736 Supplier Cayman Chemical Supplier No. 20769500 MG
A non-nitrogenous bisphosphonate that binds to hydroxyapatite and reduces osteoclast proliferation, thereby inhibiting abnormal bone resorption