Learn More
Medchemexpress LLC Etidronic acid | 2809-21-4 | 60% | 206.03 g/mol | 1 G
SDP
Supplier:  Medchemexpress LLC 1GHYB0302
Encompass_Preferred
Etidronic acid, 60% in water, is an analytical standard and an orally and intravenously active bisphosphonate. It is intended for research and analytical applications.
  • Inhibits resorption of bone
  • Reduces arterial calcification
  • Can be used for osteoporosis research
  • Has anticancer activity
  • Functions as a chelating agent to remove heavy metals in water
Catalog No. 50-003-65183