Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Etidronic acid

Catalog No. H6773100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H6773100G 100 g
H6773500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H6773100G Supplier Sigma Aldrich Supplier No. H6773100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Etidronic acid (hebp) can be employed as a catalyst for the synthesis of:
  • 5-nitro-3,4-dihydropyrimidin-2(1h)-ones through the reaction between 1-(2-hydroxyphenyl)-2-nitroethanone, aryl aldehydes and urea via one-pot cyclo condensation reaction
  • Thiiranes from epoxides and ammonium thiocyanate.tandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf. h6773.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database. to learn more, visit sigma.com/metlin

Specifications

CAS 2809-21-4
Molecular Formula C2H8O7P2
Linear Formula CH3C(OH)[PO(OH)2]2
MDL Number MFCD00070585
Quantity 100 g
Synonym 1-Hydroxyethane-1,1,-diphosphonic acid; 1-Hydroxyethylidenediphosphonic acid; HDEPA; HEDP; OEDFK
Molecular Weight (g/mol) 206.03
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.