Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Etidronic Acid, Approx. 60%, Spectrum™ Chemical
SDP

Catalog No. 18602583 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-583 100 g Amber Glass Bottle
18-602-584 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-602-583 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E3490100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

E3490, 2809-21-4, C2H8O7P2

Spectrum™ Chemical Etidronic Acid , also known as 1-hydroxyethane 1,1-diphosphonic acid, is used as a bisphosphonate to treat osteoporosis. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 2809-21-4
Molecular Formula C2H8O7P2
Molecular Weight (g/mol) 206.03
InChI Key DBVJJBKOTRCVKF-UHFFFAOYSA-N
IUPAC Name (1-hydroxy-1-phosphonoethyl)phosphonic acid
SMILES CC(O)(P(O)(O)=O)P(O)(O)=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity ∼60%
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.