Learn More
Medchemexpress LLC Etidronic acid-d3 disodium | 358730-93-5 | 98.0% | 1 MG
SDP
Catalog No. 5000405878
Encompass_Preferred
Etidronic acid-d3 disodium is a deuterium labeled compound, serving as a stable isotope-labeled compound.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.
  • Deuteration has potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-05878 Supplier Medchemexpress LLC Supplier No. HYB0302AS1MG
Etidronic acid-d3 disodium is a deuterium labeled compound, serving as a stable isotope-labeled compound.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.
  • Deuteration has potential to influence the pharmacokinetic and metabolic profiles of drugs.