Learn More
Medchemexpress LLC Etidronic acid-d3 disodium | 358730-93-5 | 98.0% | 5 MG
SDP
Catalog No. 5000407026
Encompass_Preferred
Etidronic acid-d3 disodium is a deuterium-labeled form of Etidronic acid disodium, intended exclusively for research purposes. This compound serves as a valuable tool in quantitative analysis due to the potential of deuteration to influence pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-07026 Supplier Medchemexpress LLC Supplier No. HYB0302AS5MG
Etidronic acid-d3 disodium is a deuterium-labeled form of Etidronic acid disodium, intended exclusively for research purposes. This compound serves as a valuable tool in quantitative analysis due to the potential of deuteration to influence pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs