Learn More
Description
- Etidronic acid monohydrate also known as 1-hydroxyethylidenediphosphonic acid (hedp) is an organophosphonic acid that can be used to surface functionalize nay molecular sieves
- Hedp/nay can be used as a catalyst for n-butyl acetate production via esterification.it can be also employed as a chelating agent in the synthesis of metal-ligand complexes.powerful complexing agent
Specifications
Specifications
| CAS | 25211-86-3 |
| Molecular Formula | C2H8O7P2 -+ H2O |
| Linear Formula | C2H8O7P2 -+ H2O |
| MDL Number | MFCD10567440 |
| Quantity | 50 g |
| Synonym | (1-Hydroxyethylidene)bisphosphonic acid monohydrate; 1-Hydroxyethylidenediphosphonic acid |
| Molecular Weight (g/mol) | 224.04 |
| Percent Purity | ≥95.0% (T) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.