Learn More
Medchemexpress LLC Etifenin (EHIDA) | 63245-28-3 | MFCD00059631 | 99.7% | 322.36 | C16H22N2O5 | 250 MG
SDP
Catalog No. 5000283766
Encompass_Preferred
Etifenin (EHIDA) is a small-molecule research reagent used as a precursor in the synthesis of non-toxic radiopharmaceuticals for diagnostic imaging. Intended for laboratory research use only, the compound is supplied with supporting documentation to aid safe handling and analytical confirmation.
  • High purity (99.7%).
  • Suitable precursor for radiopharmaceutical synthesis.
  • Provided with datasheet, certificate of analysis, and safety data sheet.
  • Molecular weight 322.36 and formula C16H22N2O5.
  • Available in laboratory-scale pack sizes, including 250 mg.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-83766 Supplier Medchemexpress LLC Supplier No. HYW143890250MG
Etifenin (EHIDA) is a small-molecule research reagent used as a precursor in the synthesis of non-toxic radiopharmaceuticals for diagnostic imaging. Intended for laboratory research use only, the compound is supplied with supporting documentation to aid safe handling and analytical confirmation.
  • High purity (99.7%).
  • Suitable precursor for radiopharmaceutical synthesis.
  • Provided with datasheet, certificate of analysis, and safety data sheet.
  • Molecular weight 322.36 and formula C16H22N2O5.
  • Available in laboratory-scale pack sizes, including 250 mg.
  • Intended for research use only; not for human or clinical use.