Learn More
Cayman Chemical Etifoxine-13C-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 2738376-67-3, 1 mg
SDP
Catalog No. 502990458
Encompass_Preferred
An internal standard for the quantification of etifoxine by GC- or LC-MS
Catalog No. 50-299-0458 Supplier Cayman Chemical Supplier No. 302351 MG
An internal standard for the quantification of etifoxine by GC- or LC-MS