Learn More
Cayman Chemical EtIfoxIn-13C-d3 500ug
SDP
Catalog No. 502991249
Encompass_Preferred
An internal standard for the quantification of etifoxine by GC- or LC-MS
Catalog No. 50-299-1249 Supplier Cayman Chemical Supplier No. 30235500 UG
An internal standard for the quantification of etifoxine by GC- or LC-MS