Learn More
Medchemexpress LLC Etifoxine hydrochloride | 56776-32-0 | 99.8% | 337.24 | 10 MM 1 ML
SDP
Catalog No. 5000391212
Encompass_Preferred
Etifoxine hydrochloride is a non-benzodiazepine GABAergic compound. It acts as a positive allosteric modulator of α1β2γ2 and α1β3γ2 subunit-containing GABAA receptors. It has demonstrated anxiolytic and anticonvulsant properties in rodents.
  • Acts as a positive allosteric modulator of GABAA receptors.
  • Demonstrates anxiolytic properties.
  • Demonstrates anticonvulsant properties.
Catalog No. 50-003-91212 Supplier Medchemexpress LLC Supplier No. HY1657910MM/1ML
Etifoxine hydrochloride is a non-benzodiazepine GABAergic compound. It acts as a positive allosteric modulator of α1β2γ2 and α1β3γ2 subunit-containing GABAA receptors. It has demonstrated anxiolytic and anticonvulsant properties in rodents.
  • Acts as a positive allosteric modulator of GABAA receptors.
  • Demonstrates anxiolytic properties.
  • Demonstrates anticonvulsant properties.