Learn More
Medchemexpress LLC Etifoxine (hydrochloride) | 56776-32-0 | 99.8% | 337.24 | 100 MG
SDP
Catalog No. 5000391715
Encompass_Preferred
Etifoxine hydrochloride is a non-benzodiazepine GABAergic compound that acts as a positive allosteric modulator of α1β2γ2 and α1β3γ2 subunit-containing GABAA receptors. It exhibits anxiolytic and anticonvulsant properties in rodents. This product is intended for research and analytical applications.
  • Positive allosteric modulator of GABAA receptors.
  • Exhibits anxiolytic properties.
  • Exhibits anticonvulsant properties.
  • Intended for research and analytical applications.
Catalog No. 50-003-91715 Supplier Medchemexpress LLC Supplier No. HY16579100MG
Etifoxine hydrochloride is a non-benzodiazepine GABAergic compound that acts as a positive allosteric modulator of α1β2γ2 and α1β3γ2 subunit-containing GABAA receptors. It exhibits anxiolytic and anticonvulsant properties in rodents. This product is intended for research and analytical applications.
  • Positive allosteric modulator of GABAA receptors.
  • Exhibits anxiolytic properties.
  • Exhibits anticonvulsant properties.
  • Intended for research and analytical applications.