Learn More
Medchemexpress LLC Etifoxine (hydrochloride) | 56776-32-0 | 99.8% | 337.24 | 50 MG
SDP
Catalog No. 5000392177
Encompass_Preferred
Etifoxine hydrochloride is a non-benzodiazepine GABAergic compound that acts as a positive allosteric modulator of α1β2γ2 and α1β3γ2 subunit-containing GABAA receptors, demonstrating anxiolytic and anticonvulsant properties.
  • Non-benzodiazepine GABAergic compound
  • Acts as a positive allosteric modulator of α1β2γ2 and α1β3γ2 subunit-containing GABAA receptors
  • Demonstrates anxiolytic and anticonvulsant properties
  • Increases [3H]muscimol binding
  • Competitively inhibits [35S]TBPS binding
Catalog No. 50-003-92177 Supplier Medchemexpress LLC Supplier No. HY1657950MG
Etifoxine hydrochloride is a non-benzodiazepine GABAergic compound that acts as a positive allosteric modulator of α1β2γ2 and α1β3γ2 subunit-containing GABAA receptors, demonstrating anxiolytic and anticonvulsant properties.
  • Non-benzodiazepine GABAergic compound
  • Acts as a positive allosteric modulator of α1β2γ2 and α1β3γ2 subunit-containing GABAA receptors
  • Demonstrates anxiolytic and anticonvulsant properties
  • Increases [3H]muscimol binding
  • Competitively inhibits [35S]TBPS binding