Learn More
Medchemexpress LLC Etifoxine hydrochloride | 56776-32-0 | MFCD00868190 | 99.9% | 337.24 g·mol⁻¹ | C17H18Cl2N2O | 10 MG
SDP
Catalog No. 5000289567
Encompass_Preferred
Etifoxine hydrochloride is a non-benzodiazepine GABAergic compound used in research to modulate GABAA receptor activity. It is supplied as the hydrochloride salt for improved handling and solubility, and is commonly used in preclinical studies of anxiolytic and anticonvulsant effects.
  • Acts as a positive allosteric modulator of GABAA receptors
  • Hydrochloride salt form for enhanced solubility
  • High purity suitable for research applications
  • Available in small research pack sizes for preclinical work
  • Used in studies of anxiolytic and anticonvulsant pharmacology
Catalog No. 50-002-89567 Supplier Medchemexpress LLC Supplier No. HY1657910MG
Etifoxine hydrochloride is a non-benzodiazepine GABAergic compound used in research to modulate GABAA receptor activity. It is supplied as the hydrochloride salt for improved handling and solubility, and is commonly used in preclinical studies of anxiolytic and anticonvulsant effects.
  • Acts as a positive allosteric modulator of GABAA receptors
  • Hydrochloride salt form for enhanced solubility
  • High purity suitable for research applications
  • Available in small research pack sizes for preclinical work
  • Used in studies of anxiolytic and anticonvulsant pharmacology