Learn More
Medchemexpress LLC HY-16579 5mg Medchemexpress, Etifoxine (hydrochloride) CAS:56776-32-0 Purity:>98%
SDP
Catalog No. 502081895
Encompass_Preferred
Medchemexpress, HY-16579 5mg Etifoxine (hydrochloride) CAS:56776-32-0 Etifoxine hydrochloride, a non-benzodiazepine GABAergic compound, is a positive allosteric modulator of α1β2γ2 and α1β3γ2 subunit-containing GABAA receptors. Etifoxine hydrochloride reveals anxiolytic and anticonvulsant properties in rodents. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1895 Supplier Medchemexpress LLC Supplier No. HY165795MG
Medchemexpress, HY-16579 5mg Etifoxine (hydrochloride) CAS:56776-32-0 Etifoxine hydrochloride, a non-benzodiazepine GABAergic compound, is a positive allosteric modulator of α1β2γ2 and α1β3γ2 subunit-containing GABAA receptors. Etifoxine hydrochloride reveals anxiolytic and anticonvulsant properties in rodents. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.