Learn More
Medchemexpress LLC Etilefrine | 709-55-7 | >97.03% | C10H15NO2 | 250 MG
SDP
Catalog No. 5000358022
Encompass_Preferred
Etilefrine is a sympathetic nerve agonist and AMPK activator that selectively targets α1/β1 adrenergic receptors. It enhances myocardial contractility, increases heart rate, and improves cardiac output. This compound is valuable in cardiovascular research for conditions such as postural hypotension and chylothorax.
  • Sympathetic nerve agonist and AMPK activator
  • Selectively targets α1/β1 adrenergic receptors
  • Stimulates α1 adrenergic receptors
  • Enhances myocardial contractility
  • Increases heart rate
  • Increases blood pressure
  • Regulates the AMPK/Akt pathway
  • Used in cardiovascular research
Catalog No. 50-003-58022 Supplier Medchemexpress LLC Supplier No. 250MGHYA0144
Etilefrine is a sympathetic nerve agonist and AMPK activator that selectively targets α1/β1 adrenergic receptors. It enhances myocardial contractility, increases heart rate, and improves cardiac output. This compound is valuable in cardiovascular research for conditions such as postural hypotension and chylothorax.
  • Sympathetic nerve agonist and AMPK activator
  • Selectively targets α1/β1 adrenergic receptors
  • Stimulates α1 adrenergic receptors
  • Enhances myocardial contractility
  • Increases heart rate
  • Increases blood pressure
  • Regulates the AMPK/Akt pathway
  • Used in cardiovascular research