Learn More
Medchemexpress LLC Etilefrine hydrochloride | 943-17-9 | 99.9% | C10H16ClNO2 | 100 MG
SDP
Catalog No. 5000432479
Encompass_Preferred
Etilefrine hydrochloride is a sympathetic nerve agonist and AMPK activator that selectively targets α1/β1 adrenergic receptors. It stimulates α1 adrenergic receptors, leading to contraction of vascular smooth muscle and increased peripheral resistance. It also stimulates β1 receptors to enhance myocardial contractility and increase heart rate, thereby increasing blood pressure and improving cardiac output. Etilefrine hydrochloride also bidirectionally regulates the AMPK/Akt pathway and modulates the phosphorylation levels.
  • Sympathetic nerve agonist and AMPK activator
  • Selectively targets α1/β1 adrenergic receptors
  • Increases blood pressure and improves cardiac output
  • Bidirectionally regulates the AMPK/Akt pathway
  • Applicable in cardiovascular research for conditions like postural hypotension, chylothorax, and improving low cardiac output
Catalog No. 50-004-32479 Supplier Medchemexpress LLC Supplier No. HYA0144A100MG
Etilefrine hydrochloride is a sympathetic nerve agonist and AMPK activator that selectively targets α1/β1 adrenergic receptors. It stimulates α1 adrenergic receptors, leading to contraction of vascular smooth muscle and increased peripheral resistance. It also stimulates β1 receptors to enhance myocardial contractility and increase heart rate, thereby increasing blood pressure and improving cardiac output. Etilefrine hydrochloride also bidirectionally regulates the AMPK/Akt pathway and modulates the phosphorylation levels.
  • Sympathetic nerve agonist and AMPK activator
  • Selectively targets α1/β1 adrenergic receptors
  • Increases blood pressure and improves cardiac output
  • Bidirectionally regulates the AMPK/Akt pathway
  • Applicable in cardiovascular research for conditions like postural hypotension, chylothorax, and improving low cardiac output