Learn More
Cayman Chemical Etilefrine (hydrochloride), Purity: ≥98%, CAS: 943-17-9, 1 g
SDP
Catalog No. 502950401
Encompass_Preferred
A sympathomimetic amine; induces vasoconstriction in isolated perfused rat tail artery segments in a concentration-dependent manner; inhibits anesthesia-induced decreases in mean arterial pressure and cardiac output in dogs
Catalog No. 50-295-0401 Supplier Cayman Chemical Supplier No. 332491 G
A sympathomimetic amine; induces vasoconstriction in isolated perfused rat tail artery segments in a concentration-dependent manner; inhibits anesthesia-induced decreases in mean arterial pressure and cardiac output in dogs