Learn More
Medchemexpress LLC Etilevodopa hydrochl 100mg | 39740-30-2 | 100 MG
SDP
Catalog No. 5000307673
Encompass_Preferred
Etilevodopa hydrochloride is the ethyl-ester prodrug of levodopa supplied as the hydrochloride salt for laboratory research; it is rapidly hydrolyzed to levodopa and ethanol and is used in studies of Parkinson's disease and dopamine metabolism.
  • Prodrug of levodopa with improved gastric solubility and CNS penetration.
  • Rapidly hydrolyzed to levodopa by nonspecific esterases.
  • Provided as a hydrochloride salt for enhanced stability and handling.
  • High purity suitable for research applications.
  • Available in common research pack sizes including 100 mg.
Catalog No. 50-003-07673 Supplier Medchemexpress LLC Supplier No. HY116016A100MG
Etilevodopa hydrochloride is the ethyl-ester prodrug of levodopa supplied as the hydrochloride salt for laboratory research; it is rapidly hydrolyzed to levodopa and ethanol and is used in studies of Parkinson's disease and dopamine metabolism.
  • Prodrug of levodopa with improved gastric solubility and CNS penetration.
  • Rapidly hydrolyzed to levodopa by nonspecific esterases.
  • Provided as a hydrochloride salt for enhanced stability and handling.
  • High purity suitable for research applications.
  • Available in common research pack sizes including 100 mg.