Learn More
Medchemexpress LLC Etimizol | 64-99-3 | 99.9% | 210.23 | 50 MG
SDP
Catalog No. 5000349997
Encompass_Preferred
Etimizol (Ethymisole), a nootropic agent, effectively relieves amnesia, especially when linked to hypoxic components like hypobaric hypoxia, actinomycin D, or mechanical brain injury. It works by decreasing the K-+ permeability of neuron membranes during action potential. This product is intended for research use only.
  • Effectively relieves amnesia
  • Decreases K-+ permeability of neuron membranes
  • Suitable for research applications
  • Acts as a potassium channel inhibitor
Catalog No. 50-003-49997 Supplier Medchemexpress LLC Supplier No. HY1391850MG
Etimizol (Ethymisole), a nootropic agent, effectively relieves amnesia, especially when linked to hypoxic components like hypobaric hypoxia, actinomycin D, or mechanical brain injury. It works by decreasing the K-+ permeability of neuron membranes during action potential. This product is intended for research use only.
  • Effectively relieves amnesia
  • Decreases K-+ permeability of neuron membranes
  • Suitable for research applications
  • Acts as a potassium channel inhibitor