Learn More
Medchemexpress LLC Etiocholanolone (5β-Androsterone) | 53-42-9 | 99.5% | 290.44 | 10 MG
SDP
Catalog No. 5000436460
Encompass_Preferred
Etiocholanolone (5β-Androsterone) is an excreted metabolite of testosterone with anticonvulsant activity. It is also described as a less potent neurosteroid positive allosteric modulator (PAM) of the GABAA receptor compared to its enantiomer form.
  • Excreted metabolite of testosterone
  • Anticonvulsant activity
  • Neurosteroid positive allosteric modulator (PAM) of the GABAA receptor
Catalog No. 50-004-36460 Supplier Medchemexpress LLC Supplier No. HY11332010MG
Etiocholanolone (5β-Androsterone) is an excreted metabolite of testosterone with anticonvulsant activity. It is also described as a less potent neurosteroid positive allosteric modulator (PAM) of the GABAA receptor compared to its enantiomer form.
  • Excreted metabolite of testosterone
  • Anticonvulsant activity
  • Neurosteroid positive allosteric modulator (PAM) of the GABAA receptor