Learn More
Medchemexpress LLC Etiocholanolone (5β-Androsterone) | 53-42-9 | 99.5% | 290.44 | 50 MG
SDP
Catalog No. 5000436941
Encompass_Preferred
Etiocholanolone (5β-Androsterone) is the excreted metabolite of testosterone and has anticonvulsant activity. Etiocholanolone is a less potent neurosteroid positive allosteric modulator (PAM) of the GABAA receptor than its enantiomer form.
  • It is the excreted metabolite of testosterone.
  • It has anticonvulsant activity.
  • It is a less potent neurosteroid positive allosteric modulator of the GABAA receptor than its enantiomer form.
  • It exhibits ED50 values of 57.6 and 109.1 mg/kg in the 6-Hz and PTZ tests, respectively.
Catalog No. 50-004-36941 Supplier Medchemexpress LLC Supplier No. HY11332050MG
Etiocholanolone (5β-Androsterone) is the excreted metabolite of testosterone and has anticonvulsant activity. Etiocholanolone is a less potent neurosteroid positive allosteric modulator (PAM) of the GABAA receptor than its enantiomer form.
  • It is the excreted metabolite of testosterone.
  • It has anticonvulsant activity.
  • It is a less potent neurosteroid positive allosteric modulator of the GABAA receptor than its enantiomer form.
  • It exhibits ED50 values of 57.6 and 109.1 mg/kg in the 6-Hz and PTZ tests, respectively.