Learn More
Medchemexpress LLC Etiocholanolone (5β-androsterone) | 53-42-9 | MFCD00064133 | 99.5% | 290.44 g/mol | C19H30O2 | 5 MG
SDP
Catalog No. 5000263442
Encompass_Preferred
Etiocholanolone (5β-androsterone) is an endogenous metabolite of testosterone provided as a high-purity research standard. It demonstrates anticonvulsant activity and functions as a neurosteroid positive allosteric modulator (PAM) of GABAA receptors, making it useful for biochemical, pharmacological, and analytical applications.
Catalog No. 50-002-63442 Supplier Medchemexpress LLC Supplier No. HY1133205MG
Etiocholanolone (5β-androsterone) is an endogenous metabolite of testosterone provided as a high-purity research standard. It demonstrates anticonvulsant activity and functions as a neurosteroid positive allosteric modulator (PAM) of GABAA receptors, making it useful for biochemical, pharmacological, and analytical applications.