Learn More
Cayman Chemical EtIocholndIon 10mg
SDP
Catalog No. 502949390
Encompass_Preferred
A neurosteroid; inhibits IGly in isolated rat hippocampal pyramidal neurons at 50 µM; inhibits glucose exchange transport in isolated human erythrocytes at 0.1, 0.3, and 0.5 mM
Catalog No. 50-294-9390 Supplier Cayman Chemical Supplier No. 3435810 MG
A neurosteroid; inhibits IGly in isolated rat hippocampal pyramidal neurons at 50 µM; inhibits glucose exchange transport in isolated human erythrocytes at 0.1, 0.3, and 0.5 mM