Learn More
Cayman Chemical Etiocholanolone, Purity: ≥98%, CAS: 53-42-9, 50 mg
SDP
Catalog No. 502947582
Encompass_Preferred
An androgenically inactive metabolite of testosterone and androstenedione; potentiates GABA-induced chloride currents in HEK293 cells expressing rat α1β2γ2L subunit-containing GABAA receptors at 10 µM; protective against seizures induced by 6 Hz electroshock or PTZ (ED50s = 76.9 and 139 mg/kg, respectively); increases body temperature in rhesus macaques
Catalog No. 50-294-7582 Supplier Cayman Chemical Supplier No. 3450150 MG
An androgenically inactive metabolite of testosterone and androstenedione; potentiates GABA-induced chloride currents in HEK293 cells expressing rat α1β2γ2L subunit-containing GABAA receptors at 10 µM; protective against seizures induced by 6 Hz electroshock or PTZ (ED50s = 76.9 and 139 mg/kg, respectively); increases body temperature in rhesus macaques