Learn More
Medchemexpress LLC HY-B0106A 50mg Medchemexpress, Etiracetam CAS:33996-58-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0106A50MG
Encompass_Preferred
Medchemexpress, HY-B0106A 50mg Etiracetam CAS:33996-58-6 Etiracetam (UCB 6474) is an acetylcholine agonist and a nootropic drug of the racetam family. Less active than its S-enantiomer Levetiracetam (UCB L059). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0097